missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Growth Hormone 2 Monoclonal antibody specifically detects Growth Hormone 2 in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
Specifications
| Antigen | Growth Hormone 2 |
| Applications | Western Blot, ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 1E11 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Sandwich ELISA |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | AAH20760 |
| Gene Alias | GHL, GHVgrowth hormone variant, GH-VPlacenta-specific growth hormone, growth hormone 2hGH-V, placental-specific growth hormone |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?