missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ground& Waste Water Pollution Standard Solution 5, Specpure™

Brand:  Alfa Aesar™ 042580.AE

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Quantity 100mL

 View more versions of this product

Product Code. 11339198

  • £93.90 / 100mL


Estimated Shipment date
from Supplier 25-06-2018
Add to basket

Specifications

Chemical Identifiers

Ground & Waste Water Pollution Standard Solution 5
Specpure™
MFCD03792307
UN3264

Specifications

Ground & Waste Water Pollution Standard Solution 5
Liquid
UN3264
Matrix: 2% HNO3
Specpure™
MFCD03792307
100mL
Videos
SDS