missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ground& Waste Water Pollution Standard Solution 4, Specpure™

Brand:  Alfa Aesar™ 042579.AE

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Quantity 100mL

 View more versions of this product

Product Code. 11329198

  • £149.40 / 100mL


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Specifications

Chemical Identifiers

Ground & Waste Water Pollution Standard Solution 4
Specpure™
MFCD03792306
UN3264

Specifications

Ground & Waste Water Pollution Standard Solution 4
Liquid
UN3264
Matrix: 2% HNO3/tr. tartaric acid
Specpure™
MFCD03792306
100mL
Videos
SDS