missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
- This item is an accessory part for the Millipore All-Glass Filter Holders Kits and Millipore MilliSolve Kit.
Specifications
Specifications
| Color | Clear |
| Disposable | Reusable |
| For Use With (Equipment) | Millipore All-glass Filter Holders Kit, Millipore MilliSolve Kit |
| Material | Borosilicate Glass |
| Shape | Conical Bottom |
| Type | Conical Bottom Flask |
| Volume (Metric) | 1 L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?