missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ groL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144463
This item is not returnable.
View return policy
Description
groL Polyclonal Antibody for Western Blot, ELISA
Together with its co-chaperonin GroES, plays an essential role in assisting protein folding. The GroEL-GroES system forms a nano-cage that allows encapsulation of the non-native substrate proteins and provides a physical environment optimized to promote and accelerate protein folding.
Specifications
| groL | |
| Polyclonal | |
| Unconjugated | |
| groEL | |
| groL; htpB; mopA | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 1209629 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3.07 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P19421 | |
| groEL | |
| Recombinant Coxiella burnetii (strain RSA 493/Nine Mile phase I) CH60 protein. | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction