missing translation for 'onlineSavingsMsg'
Learn More

GRO2 Antibody (RM0209-13N6), DyLight 594, Novus Biologicals™

Product Code. 18401627 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401627 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401627 Supplier Novus Biologicals Supplier No. NBP212228DL594

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rat Monoclonal Antibody

GRO2 Monoclonal antibody specifically detects GRO2 in Mouse samples. It is validated for Western Blot, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRO2
Applications Western Blot, Neutralization
Classification Monoclonal
Clone RM0209-13N6
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias chemokine (C-X-C motif) ligand 2, CINC-2a, GRO2 oncogene, GRO2MIP2, GROb, gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MIP-2a, MIP2AC-X-C motif chemokine 2, SCYB2MIP2-alpha
Host Species Rat
Immunogen Mouse recombinant GRO2
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2920
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.