missing translation for 'onlineSavingsMsg'
Learn More

GRO Hydrocarbons Mixture (C4-C12) 631 2000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30308765
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30308765

Brand: Dr. Ehrenstorfer DREA50000631ME

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Butane∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Nonane∣n-Octane∣n-Pentane∣n-Undecane
CAS 106-97-8, 124-18-5, 112-40-3, 142-82-5, 110-54-3, 111-84-2
Analyte or Component Names n-Butane∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Nonane∣n-Octane∣n-Pentane∣n-Undecane
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) n-Butane 2000 μg/mL∣n-Decane 2000 μg/mL∣n-Dodecane 2000 μg/mL∣n-Heptane 2000 μg/mL∣n-Hexane 2000 μg/mL∣n-Nonane 2000 μg/mL∣n-Octane 2000 μg/mL∣n-Pentane 2000 μg/mL∣n-Undecane 2000 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.