missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRM8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18224-1-AP-150UL
This item is not returnable.
View return policy
Description
L-glutamate is the major excitatory neurotransmitter in the central nervous system and activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors, that have been divided into 3 groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5 and these receptors have been shown to activate phospholipase C. Group II includes GRM2 and GRM3 while Group III includes GRM4, GRM6, GRM7 and GRM8. Group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities.Specifications
| GRM8 | |
| Polyclonal | |
| Unconjugated | |
| GRM8 | |
| GLUR8, GPRC1H, GRM8, mGlu8, MGLUR8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14823, 2918, 60590 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00222, P47743, P70579 | |
| Grm8 | |
| GRM8 Fusion Protein Ag13011 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction