missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRK7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA514807
This item is not returnable.
View return policy
Description
GRK7 Polyclonal Antibody for Western Blot, IHC (P)
GRK7 is a serine/threonine kinase that is specifically expressed in the retina and phosphorylates cone opsins and initiates their deactivation. Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e.g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e.g. ABL, FAK, JAK, and SRC families).
Specifications
| GRK7 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8WTQ7 | |
| GRK7 | |
| KLH conjugated synthetic peptide between 508-538 amino acids from the C-terminal region of human GRK7. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| GRK7 | |
| G protein-coupled receptor kinase 7; g protein-coupled receptor kinase GRK7; GPRK7; GRK7; rhodopsin kinase; Rhodopsin kinase GRK7 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 131890 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction