missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRK6 Recombinant Rabbit Monoclonal Antibody (K01_3L85)
Description
Immunogen sequence: KRKACGRQLT QNFL.
GRK6 is a member of the guanine nucleotide-binding protein coupled receptor kinase subfamily of the serine/threonine protein kinase family. GRK6 phosphorylates the activated forms of G protein-coupled receptors to initiate their deactivation. Several transcript variants encoding different isoforms have been described for GRK6.
Specifications
Specifications
| Antigen | GRK6 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_3L85 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | Grk6 |
| Gene Accession No. | O70293, P43250 |
| Gene Alias | activin receptor IIA; activin receptor type IIA; Activin receptor type-2A; ActrIIa; ACTR-IIA; Acvr2; Acvr2a; EC 2.7.11.16; G protein-coupled receptor kinase 6; g protein-coupled receptor kinase GRK6; Gprk6; G-protein coupled receptor kinase 6; GRK6; kinase GRK6; TactrII |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?