Learn More
Invitrogen™ GRK6 Recombinant Human Protein
GRK6 is a member of the guanine nucleotide-binding protein coupled receptor kinase subfamily of the serine/threonine protein kinase family.
Brand: Invitrogen™ PR6302C
Code : NEW
Additional Details : Weight : 0.01000kg
Description
GRK6 phosphorylates the activated forms of G protein-coupled receptors to initiate their deactivation.
Specifications
NP_001004106 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
GRK6, GPRK6 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
94kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
GRK Kinase Family | |
Recombinant | |
Baculovirus |