missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK6 Antibody (S07-3E4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19869-100ul
This item is not returnable.
View return policy
Description
GRK6 Monoclonal antibody specifically detects GRK6 in Human, Mouse samples. It is validated for Western Blot
Specifications
| GRK6 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| S07-3E4 | |
| Unconjugated | |
| EC 2.7.11, EC 2.7.11.16, FLJ32135, G protein-coupled receptor kinase 6, GPRK6G protein-coupled receptor kinase GRK6 | |
| A synthetic peptide of human GRK6 (Uniprot # P43250) | |
| 100 μg | |
| Protein Kinase | |
| 2870 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction