missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13727-1-AP-20UL
This item is not returnable.
View return policy
Description
The beta-adrenergic receptor kinase specifically phosphorylates the agonist-occupied form of the beta-adrenergic and related G protein-coupled receptors. Overall, the ADRBK2 enzyme, also known as GRK3, has 85% amino acid similarity with ADRBK1, with the protein kinase catalytic domain having 95% similarity. The ADRBK2 mRNA is approximately 8 kilobases with a distribution similar to that of ADRBK1. These data suggest the existence of a family of receptor kinases which may serve broadly to regulate receptor function.Specifications
| GRK3 | |
| Polyclonal | |
| Unconjugated | |
| GRK3 | |
| ADRBK2, BARK2, Beta ARK 2, GRK3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 157, 25372, 320129 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P26819, P35626 | |
| Grk3 | |
| GRK3 Fusion Protein Ag4685 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction