missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRK3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16358864
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16358864 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16358864 Supplier Invitrogen™ Supplier No. PA581896

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: YEAVNADTDK IEARKRAKNK QLGHEEDYAL GKDCIMHGYM LKLGNPFLTQ WQRRYFYLFP NRLEWRGEGE SRQNLLTMEQ ILSVEETQIK DKKCILFRIK GGKQFVLQCE SDPEFVQWKK ELNETFKEAQ RLLRRAPKFL NKPRS Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 89%.

ADRBK2, or adrenergic beta receptor kinase 2 is a serine/threonine kinase that is thought to act as a regulator of receptor function. Overall, the ADRBK2 enzyme, also known as GRK3, has 85% amino acid similarity with ADRBK1, with the protein kinase catalytic domain having 95% similarity. The ADRBK2 mRNA is approximately 8 kilobases with a distribution similar to that of ADRBK1. These data suggest the existence of a family of receptor kinases which may serve broadly to regulate receptor function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRK3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRK3
Gene Accession No. P35626
Gene Alias 4833444A01Rik; ADRBK2; Adrbk-2; adrenergic receptor kinase, beta 2; adrenergic receptor kinase, beta 2 (G-protein-linked receptor kinase); adrenergic, beta, receptor kinase 2; AI851927; AW551196; BARK2; Bark-2; beta ARK2; beta-adrenergic receptor kinase 2; Beta-ARK-2; G protein-coupled receptor kinase 3; G-protein-coupled receptor kinase 3; Grk3
Gene Symbols GRK3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GRK3. Recombinant protein control fragment (Product #RP-91358).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.