missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103041
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GRK1.
GRK1 is a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinases. GRK1 mediates rapid desensitization of rod photoreceptors to light by catalyzing phosphorylation of the visual pigment rhodopsin which leads to its deactivation. Defects in GRK1 are known to cause Oguchi disease 2 (also known as stationary night blindness Oguchi type-2). GRK1 can be phosphorylated by PKA and this reduces the ability of GRK1 to phosphorylate rhodopsin in vitro. It is speculated that phosphorylation of GRK1 by PKA in vivo occurs in the dark when cAMP levels in photoreceptor cells are elevated.
Specifications
| GRK1 | |
| Polyclonal | |
| Unconjugated | |
| GRK1 | |
| G protein-coupled receptor kinase 1; G protein-coupled receptpr kinase 1; GPRK1; G-protein coupled receptor kinase 1; G-protein receptor kinase 1; GRK 1a/b; GRK1; GRK1a; rhodopsin kinase; rhodopsin kinase GRK1; Rhok; RK | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 24013, 6011, 81760 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15835, Q63651, Q9WVL4 | |
| GRK1 | |
| A synthesized peptide derived from human GRK1(Accession Q15835), corresponding to amino acid residues M1-K46. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction