missing translation for 'onlineSavingsMsg'
Learn More

Griseofulvin, TRC

Product Code. 30400963
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30400963 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400963 Supplier Toronto Research Chemicals Supplier No. TRCG78750050G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Griseofulvine
CAS 126-07-8
Purity Grade Notes HPLC
Molecular Formula C17 H17 Cl O6
Quantity 50 g
Synonym Griseofulvin, Spiro[benzofuran-2(3H),1'-[2]cyclohexene]-3,4'-dione, 7-chloro-2',4,6-trimethoxy-6'-methyl-, (1'S,6'R)-, Spiro[benzofuran-2(3H),1'-[2]cyclohexene]-3,4'-dione, 7-chloro-2',4,6-trimethoxy-6'-methyl-, (1'S-trans)-, Spiro[benzofuran-2(3H),1'-[2]cyclohexene]-3,4'-dione, 7-chloro-2',4,6-trimethoxy-6'β-methyl- (8CI), (1'S,6'R)-7-Chloro-2',4,6-trimethoxy-6'-methylspiro[benzofuran-2(3H),1'-[2]cyclohexene]-3,4'-dione, (+)-Griseofulvin, 7-Chloro-2',4,6-trimethoxy-6'β-methylspiro[benzofuran-2(3H),1'-[2]cyclohexene]-3,4'-dione, Amudane, Curling factor, Fulcin, Fulvicin, Fulvicin Bolus, Fulvicin P/G, Fulvicin U/F, Fulvidex, Fulvina, Fulvinil, Fulvistatin, Gricin, Grifulin, Grifulvin, Grifulvin V, Gris-PEG, Grisactin, Grisefulin, (+)-, Griseovin, Grisofulvin, Grisovin, Grisovin FP, Grysio, Idifulvin, Lamoryl, Likuden, Neo-Fulcin, Polygris, Poncyl, Poncyl FP, Spirofulvin, Sporostatin, Xuanjing
InChI Formula InChI=1S/C17H17ClO6/c1-8-5-9(19)6-12(23-4)17(8)16(20)13-10(21-2)7-11(22-3)14(18)15(13)24-17/h6-8H,5H2,1-4H3/t8-,17+/m1/s1
SMILES COC1=CC(=O)C[C@@H](C)[C@]12Oc3c(Cl)c(OC)cc(OC)c3C2=O
IUPAC Name (2S,5'R)-7-chloro-3',4,6-trimethoxy-5'-methylspiro[1-benzofuran-2,4'-cyclohex-2-ene]-1',3-dione
Molecular Weight (g/mol) 352.77
Formula Weight 352.0714
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.