missing translation for 'onlineSavingsMsg'
Learn More

Griseofulvin-13C,d3, TRC

Product Code. 30394017
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394017 1 mg -
30398939 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30394017 Supplier Toronto Research Chemicals Supplier No. TRCG7875021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1329612-29-4
Molecular Formula C1613CH14D3ClO6
Molecular Weight (g/mol) 356.78
Synonym (1'S,6'R)-7-Chloro-2',6-dimethoxy-4-(methoxy-d3)-6'-methyl-spiro[benzofuran-2(3H),1'-[2]cyclohexene]-3,4'-dione, Amudane-13C,d3, Fulcin-13C,d3, Grisovin-13C,d3, Spirofulvin-13C,d3, Sporostatin-13C,d3, Xuanjing-13C,d3
SMILES O=C1[C@@]2([C@H](C)CC(C=C2OC)=O)OC3=C(Cl)C(OC)=CC(O[13C]([2H])([2H])[2H])=C31

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C17H17ClO6/c1-8-5-9(19)6-12(23-4)17(8)16(20)13-10(21-2)7-11(22-3)14(18)15(13)24-17/h6-8H,5H2,1-4H3/t8-,17+/m1/s1/i2+1D3
Formula Weight 356.0936
Chemical Name or Material Griseofulvin-13C,d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.