missing translation for 'onlineSavingsMsg'
Learn More

GRIPAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen GRIPAP1
Concentration 0.16 mg/mL
Applications Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16875154
View Documents
Proteintech
15806-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16865154
View Documents
Proteintech
15806-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The MW of this protein is 97 kDa, and 15806-1-AP specifically recognizes the 97 kDa protein.

GRASP1 is a neuron-specific guanine nucleotide exchange factor for the Ras family of small G proteins (RasGEF) and is associated with the GRIP/AMPA receptor complex in brain.
Specifications

Specifications

GRIPAP1
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
Unconjugated
Rabbit
Rat, Human, Mouse
Q4V328, Q8VD04, Q9JHZ4
116493, 54645, 56850
GRIPAP1 Fusion Protein Ag8540
Primary
-20°C
GRIPAP1
0.16 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
GRASP 1, GRIP1 associated protein 1, GRIPAP1, KIAA1167
Gripap1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.