Learn More
Invitrogen™ GRIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596412
Description
Positive Samples: 293T; Cellular Location: Cell junction, Cytoplasmic vesicle, Endoplasmic reticulum, postsynaptic cell membrane, synapse Immunogen sequence: MIAVSFKCRC QILRRLTKDE SPYTKSASQT KPPDGALAVR RQSIPEEFKG STVVELMKKE GTTLGLTVSG GIDKDGKPRV SNLRQGGIAA RSDQLDVGDY IKAVNGINLA KFRHDEIISL LKNVGERVVL EVEYELPPVS VQGSSVIFRT VEVTLHKEGN TFGFVIRGGA HDDRNKSRPV VITCVRPGGP ADREGTIKPG DRLLSVDGIR LLGTTHAEAM SILKQCGQEA ALLIEYDVSV MDSVATASGP.
Steroid and thyroid hormones and retinoic acid regulate a complex array of gene expression activity via intracellular receptor transcription factors belonging to the ligand-dependent nuclear receptor superfamily. Adding to the complexity of function of these transcription factors are associated proteins known as coactivators and corepressors which, as their names suggest, enhance or depress transcription activity of the nuclear receptor with which they associate. Glucocorticoid receptor interacting protein-1 (GRIP1), or Transcription Intermediary Factor 2 (TIF2), is a member of a family of transcriptional coactivator proteins which includes steroid receptor coactivator-1 (SRC-1) and Amplified in breast cancer-1 (AIB-1). GRIP1 has been shown to interact and stimulate transcriptional activity of the retinoic acid, retinoid X, vitamin D, mineralocorticoid, glucocorticoid, thyroid hormone, androgen, estrogen, progesterone, and peroxisome proliferator activated receptors. GRIP1 also contains several copies of the conserved LXXLL/LLXXL motif which has been demonstrated to be critical to receptor-coactivator interactions.
Specifications
| GRIP1 | |
| Polyclonal | |
| Unconjugated | |
| GRIP1 | |
| 4931400F03Rik; AMPA receptor-interacting protein GRIP1; eb; eye blebs; glutamate receptor interacting protein 1; glutamate receptor-interacting protein 1; GRIP; Grip1; GRIP-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23426, 74053 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q925T6, Q9Y3R0 | |
| GRIP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human GRIP1 (NP_0011715451). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.