missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRIP1 Polyclonal Antibody

GRIP1 Polyclonal Antibody
Brand: Thermo Scientific PA5114743
This item is not returnable.
View return policy
Description
GRIP1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
Specifications
| GRIP1 | |
| Polyclonal | |
| Unconjugated | |
| GRIP1 | |
| 4931400F03Rik; AMPA receptor-interacting protein GRIP1; eb; eye blebs; glutamate receptor interacting protein 1; glutamate receptor-interacting protein 1; GRIP; Grip1; GRIP-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23426, 74053, 84016 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P97879, Q925T6, Q9Y3R0 | |
| GRIP1 | |
| A synthesized peptide derived from human GRIP1(Accession Q9Y3R0), corresponding to amino acid residues V860-L910. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction