missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRIP1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-2842AF488
This item is not returnable.
View return policy
Description
GRIP1 Polyclonal antibody specifically detects GRIP1 in Human, Hamster samples. It is validated for Immunofluorescence
Specifications
| GRIP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Hamster | |
| Purified |
| Immunofluorescence | |
| Alexa Fluor 488 | |
| glutamate receptor interacting protein 1, glutamate receptor-interacting protein 1, GRIP, GRIP-1 | |
| Synthetic peptide corresponding to residues P(206) L P D S E E E G H D S(217) C of human GRIP1. | |
| 0.1 mL | |
| Cancer, Neuronal Cell Markers, Neurotransmission, Transcription Factors and Regulators | |
| 23426 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction