Learn More
Honeywell™ GRIP Latex Gloves
Product Code. 12625203
Change view
Click to view available options
Size:
9
10
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12625203 10 Each
12655203 9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12625203 Supplier Honeywell™ Supplier No. 209414510
Only null left
Add to basket
Add to basket
Honeywell™ GRIP Latex Gloves for handling of heavy and slightly sharp parts in a damp or non greasy environment.
  • Natural latex coated glove on cotton jersey base
  • Crinkled finishing
  • Knitted wrist
  • The cotton absorbs perspiration thus provides greater comfort
  • The ventilated back improves the glove's breathability
  • The glove's crinkled latex coating provides resistance to abrasion and perforation as well as an excellent grip
  • Natural latex provides excellent flexibility, synonymous with dexterity and comfort
  • It also offers good abrasion and tear resistance
  • Its crepe finish helps the user to grasp slippery parts
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-2003 Basic standard, EN 388-2016 Mechanical risks
Coating Material Latex
Exterior Finish Crinkle
Material Cotton
Size 10
Length (Metric) 26 cm