missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRIN2B Mouse anti-Human, Clone:6E9A8, Abnova™
Mouse monoclonal antibody raised against partial recombinant human GRIN2B.
Brand: Abnova MAB17961.100ug
This item is not returnable.
View return policy
Description
N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate receptors. NMDA receptor channel has been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of three different subunits: NR1 (GRIN1), NR2 (GRIN2A, GRIN2B, GRIN2C, or GRIN2D) and NR3 (GRIN3A or GRIN3B). The NR2 subunit acts as the agonist binding site for glutamate. This receptor is the predominant excitatory neurotransmitter receptor in the mammalian brain. [provided by RefSeq]Specifications
| GRIN2B | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunofluorescence (1:100-1:500) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
| GRIN2B | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunofluorescence, Western Blot | |
| 6E9A8 | |
| glutamate receptor, ionotropic, N-methyl D-aspartate 2B | |
| In PBS (0.05% sodium azide) | |
| MGC142178, MGC142180, NMDAR2B, NR2B, hNR3 | |
| Recombinant protein corresponding to amino acids 27-163 of human GRIN2B. | |
| 2904 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction