missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRIM19 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£188.00 - £375.00
Specifications
| Antigen | GRIM19 |
|---|---|
| Applications | Western Blot, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18303216
|
Novus Biologicals
NBP3-03755-20UL |
20 μL |
£188.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18382014
|
Novus Biologicals
NBP3-03755-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRIM19 Polyclonal specifically detects GRIM19 in Human, Mouse samples. It is validated for Western Blot, Knockout Validated.Specifications
| GRIM19 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| B16.6, CDA016, Cell death regulatory protein GRIM-19, cell death-regulatory protein GRIM19, CI-B16.6, complex I B16.6 subunit, Complex I-B16.6, FLJ58045, FLJ59191, Gene associated with retinoic and IFN-induced mortality 19 protein, Gene associated with retinoic and interferon-induced mortality 19 protein, GRIM-19CGI-39, GRIM19NADH-ubiquinone oxidoreductase B16.6 subunit, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 52-144 of human GRIM19 (NP_057049.5). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer | |
| PBS with 50% glycerol, pH7.3. | |
| 51079 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title