missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GRIM19 Polyclonal specifically detects GRIM19 in Human, Mouse samples. It is validated for Western Blot, Knockout Validated.
Specifications
Specifications
| Antigen | GRIM19 |
| Applications | Western Blot, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | B16.6, CDA016, Cell death regulatory protein GRIM-19, cell death-regulatory protein GRIM19, CI-B16.6, complex I B16.6 subunit, Complex I-B16.6, FLJ58045, FLJ59191, Gene associated with retinoic and IFN-induced mortality 19 protein, Gene associated with retinoic and interferon-induced mortality 19 protein, GRIM-19CGI-39, GRIM19NADH-ubiquinone oxidoreductase B16.6 subunit, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 52-144 of human GRIM19 (NP_057049.5). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?