missing translation for 'onlineSavingsMsg'
Learn More

GRIM19 Rabbit anti-Human, Clone: 7D1Z6, Novus Biologicals™

Product Code. 18335674 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335674 100 μg 100µL
18345614 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18335674 Supplier Novus Biologicals Supplier No. NBP316274100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GRIM19 Monoclonal antibody specifically detects GRIM19 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRIM19
Applications Western Blot
Classification Monoclonal
Clone 7D1Z6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias B16.6, CDA016, Cell death regulatory protein GRIM-19, cell death-regulatory protein GRIM19, CI-B16.6, complex I B16.6 subunit, Complex I-B16.6, FLJ58045, FLJ59191, Gene associated with retinoic and IFN-induced mortality 19 protein, Gene associated with retinoic and interferon-induced mortality 19 protein, GRIM-19CGI-39, GRIM19NADH-ubiquinone oxidoreductase B16.6 subunit, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human GRIM19 (Q9P0J0).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 51079
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.