missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRIK5 Polyclonal Antibody

Product Code. 12828632
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12828632 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12828632 Supplier Invitrogen™ Supplier No. PA520451

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-20451 can be used with blocking peptide PEP-0568.

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. Grik5, also known as kainate-preferring glutamate recptor subunit KA2, belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. Grik5 is highly homologous to the related ionotrophic glutamate receptor Grik4 (also known as KA1). Like Grik4, Grik5 does not form homomeric channels, but instead forms heteromers with Grik2. In Grik2- but not Grik1-null mice, Grik5 surface expression is greatly reduced in neurons, indicating that Grik2/Grik5 heteromers are required for exit from the endoplasmic reticulum to the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRIK5
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene GRIK5
Gene Accession No. Q16478, Q61626, Q63273
Gene Alias EAA2; Excitatory amino acid receptor 2; GluK5; GluR gamma-2; GluRgamma2; glutamate ionotropic receptor kainate type subunit 5; glutamate receptor; glutamate receptor gamma-2; Glutamate receptor ionotropic, kainate 5; glutamate receptor KA2; glutamate receptor KA-2; glutamate receptor, ionotropic kainate 5; glutamate receptor, ionotropic, kainate 5; glutamate receptor, ionotropic, kainate 5 (gamma 2); GRIK2; GRIK5; iGlu5; KA2; LOW QUALITY PROTEIN: glutamate receptor ionotropic, kainate 5; MGC128958 protein
Gene Symbols GRIK5
Host Species Rabbit
Immunogen A 17 amino acid peptide near the carboxy terminus of the human Grik5.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14809, 24407, 2901
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.