missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRIK4 Polyclonal Antibody

Product Code. 12838632
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12838632 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12838632 Supplier Invitrogen™ Supplier No. PA520453

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Despite its predicted molecular weight, Grik4 often migrates at a lower molecular weight in SDS-PAGE. A suggested positive control is rat brain tissue lysate. PA5-20453 can be used with blocking peptide PEP-0570.

This gene encodes a protein that belongs to the glutamate-gated ionic channel family. Glutamate functions as the major excitatory neurotransmitter in the central nervous system through activation of ligand-gated ion channels and G protein-coupled membrane receptors. The protein encoded by this gene forms functional heteromeric kainate-preferring ionic channels with the subunits encoded by related gene family members.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRIK4
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene GRIK4
Gene Accession No. Q01812, Q16099, Q8BMF5
Gene Alias 6330551K01Rik; EAA1; Excitatory amino acid receptor 1; GluK4; GluRgamma1; glutamate ionotropic receptor kainate type subunit 4; glutamate receptor; glutamate receptor ionotropic, kainate 4; glutamate receptor KA1; Glutamate receptor KA-1; glutamate receptor, ionotropic, kainate 4; GRIK; Grik4; KA1; KA-1; kainate receceptor
Gene Symbols GRIK4
Host Species Rabbit
Immunogen A 14 amino acid peptide near the amino terminus of the human Grik4.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110637, 24406, 2900
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.