missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Griffonia Simplicifolia Lectin I (GSL I) Isolectin B4 Native Protein
Shop All Bio Techne Products
Click to view available options
:
0.5 mg
Description
- A biologically active protein corresponding to Griffonia Simplicifolia Lectin I (GSL I) Isolectin B4
- Source: Griffonia (Bandeiraea) simplicifolia seeds
- GSL I-B4 isolectin contains only the B subunits.
- It is a useful marker for endothelial cells from nonprimates such as mouse, rat, rabbit, and goat as well as a marker for non-peptidergic unmyelinated primary afferent neurons.
- This “B”-rich lectin preferentially agglutinates blood group B cells and is specific for alpha-galactose residues.
- DyLight™ 649 GSL I-B4 isolectin is produced by using special conjugation procedures to incorporate DyLight™ 649 into our affinity-purified lectin.
- This conjugate has an appropriate number of fluorochromes bound which provide the maximum fluorescence and optimum staining characteristics for this particular lectin.
- This reagent is supplied essentially free of unconjugated fluorochromes and inactive lectin.
- Excitation maximum is at 646 nm and emission maximum is at 674 nm (red).
Specifications
Specifications
| For Use With (Application) | Bioactivity, Immunocytochemistry/Immunofluorescence |
| Molecular Weight (g/mol) | 114 kDa |
| Purification Method | Affinity purified |
| Quantity | 0.5 mg |
| Storage Requirements | Store at 4°C in the dark. |
| Conjugate | DyLight 649 |
| Buffer | 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.1 mM Ca2+ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction