missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRIA4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23350-1-AP-20UL
This item is not returnable.
View return policy
Description
Glutamate receptors mediate most excitatory neurotransmission in the brain and play an important role in neural plasticity, neural development and neurodegeneration. Ionotropic glutamate receptors are categorized into NMDA receptors and kainate/AMPA receptors, both of which contain glutamate-gated, cation-specific ion channels. Kainate/AMPA receptors are co-localized with NMDA receptors in many synapses and consist of seven structurally related subunits designated GluR-1 to -7. The kainate/AMPA receptors are primarily responsible for the fast excitatory neuro-transmission by glutamate, whereas the NMDA receptors are functionally characterized by a slow kinetic and a high permeability for Ca2+ ions.Specifications
| GRIA4 | |
| Polyclonal | |
| Unconjugated | |
| GRIA4 | |
| GluA4, GluR 4, GluR D, GLUR4, GLUR4C, GLURD, Glutamate receptor 4, GRIA4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14802, 2893, 29629 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P19493, P48058, Q9Z2W8 | |
| GRIA4 | |
| GRIA4 Fusion Protein Ag19949 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction