missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRHPR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 51013-2-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes an enzyme with hydroxypyruvate reductase, glyoxylate reductase, and D-glycerate dehydrogenase enzymatic activities. The enzyme has widespread tissue expression and has a role in metabolism. Type II hyperoxaluria is caused by mutations in this gene.Specifications
| GRHPR | |
| Polyclonal | |
| Unconjugated | |
| GRHPR | |
| GLXR, GLYD, GRHPR, PH2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 76238, 9380 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q91Z53, Q9UBQ7 | |
| GRHPR | |
| GRHPR Fusion Protein Ag0484 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto