missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRHPR Monoclonal Antibody (24GB6270)

Mouse Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | GRHPR |
|---|---|
| Clone | 24GB6270 |
| Content And Storage | -20°C |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275055
![]() |
Invitrogen™
MA557416 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275313
|
Invitrogen™
MA557417 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRHPR Monoclonal Antibody for Western Blot
This gene encodes an enzyme with hydroxypyruvate reductase, glyoxylate reductase, and D-glycerate dehydrogenase enzymatic activities. The enzyme has widespread tissue expression and has a role in metabolism. Type II hyperoxaluria is caused by mutations in this gene.Specifications
| GRHPR | |
| -20°C | |
| Monoclonal | |
| Liquid | |
| IgG2a | |
| Human | |
| Grhpr | |
| 1110059D05Rik; 6430629L09Rik; Glxr; glycerate-2-dehydrogenase; GLYD; glyoxylate and hydroxypyruvate reductase; glyoxylate reductase/hydroxypyruvate reductase; GRHPR; MSTP035; PH2 | |
| Grhpr | |
| Primary | |
| Affinity chromatography |
| 24GB6270 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UBQ7 | |
| 9380 | |
| Recombinant protein of human GRHPR (aa 1-328). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title