missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRHL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541616
This item is not returnable.
View return policy
Description
Peptide sequence: EEVFDALMLK TPDLKGLRNA ISEKYGFPEE NIYKVYKKCK RGILVNMDNN Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 100%; Rat: 93%.
This gene encodes a member of the grainyhead family of transcription factors. The encoded protein interacts with leader-binding protein 32 (LBP-32) and brother of mammalian grainyhead (BOM), and may function as a transcription factor during development. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Additional transcript variants have been described, but their biological nature has not been determined.
Specifications
| GRHL3 | |
| Polyclonal | |
| Unconjugated | |
| GRHL3 | |
| AI561912; ct; Get1; grainy head-like 3; grainyhead like transcription factor 3; grainyhead-like 3; grainyhead-like 3 (Drosophila); grainyhead-like protein 3 homolog; Grhl3; Sister of mammalian grainyhead; sister-of-mammalian grainyhead; sister-of-mammalian grainyhead protein; SOM; TFCP2L4; Transcription factor CP2-like 4; transcription factor hSOM1; VWS2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57822 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TE85 | |
| GRHL3 | |
| Synthetic peptide directed towards the C-terminal of human GRHL3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction