missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GRHL2 Polyclonal antibody specifically detects GRHL2 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | GRHL2 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA |
| Formulation | 50% Glycerol |
| Gene Accession No. | NP_079191 |
| Gene Alias | BOMMGC149294, Brother of mammalian grainyhead, deafness, autosomal dominant 28, DFNA28, FLJ13782, grainyhead-like 2 (Drosophila), grainyhead-like protein 2 homolog, TFCP2L3MGC149295, Transcription factor CP2-like 3FLJ11172 |
| Host Species | Mouse |
| Immunogen | GRHL2 (NP_079191, 111 a.a. - 210 a.a.) partial recombinant protein with GST tag. ENRVQVLKTVPVNLSLNQDHLENSKREQYSISFPESSAIIPVSGITVVKAEDFTPVFMAPPVHYPRGDGEEQRVVIFEQTQYDVPSLATHSAYLKDDQRS |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?