missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRHL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16745
This item is not returnable.
View return policy
Description
GRHL2 Polyclonal specifically detects GRHL2 in Human samples. It is validated for Western Blot.
Specifications
| GRHL2 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| BOMMGC149294, Brother of mammalian grainyhead, deafness, autosomal dominant 28, DFNA28, FLJ13782, grainyhead-like 2 (Drosophila), grainyhead-like protein 2 homolog, TFCP2L3MGC149295, Transcription factor CP2-like 3FLJ11172 | |
| Rabbit | |
| 71 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (90%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.94mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q6ISB3 | |
| GRHL2 | |
| Recombinant protein encompassing a sequence within the center region of human GRHL2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 79977 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction