missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRHL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17644-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the grainyhead family of transcription factors. The encoded protein can exist as a homodimer or can form heterodimers with sister-of-mammalian grainyhead or brother-of-mammalian grainyhead. This protein functions as a transcription factor during development.Specifications
| GRHL1 | |
| Polyclonal | |
| Unconjugated | |
| GRHL1 | |
| grainyhead like 1 (Drosophila), GRHL1, LBP 32, LBP32, Mammalian grainyhead, MGR, NH32, TFCP2L2, Transcription factor LBP 32 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 195733, 29841, 313993 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q921D9, Q9NZI5 | |
| GRHL1 | |
| GRHL1 Fusion Protein Ag11832 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction