missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress GRGDSPK

Product Code. 16419488
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16419488 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16419488 Supplier MedChemExpress Supplier No. HYP0322

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GRGDSPK (EMD 56574) is a peptide incluing Arg-Gly-Asp (RGD). GRGDSPK (EMD 56574) is an competitive and reversible inhibitory peptide for inhibiting integrin-fibronectin binding. GRGDSPK is used to study the role of integrins in bone formation and resorption.

Specifications

Purity 98.3%
CAS 111119-28-9
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) COVID-19-immunoregulation
Molecular Weight (g/mol) 715.76 g/mol
Product Type Peptides
Quantity 10 mg
Shipping Condition Room temperature
Synonym EMD 56574
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.