missing translation for 'onlineSavingsMsg'
Learn More

GRG (Groucho homolog) Antibody, DyLight 350, Novus Biologicals™

Product Code. 18407665 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407665 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407665 Supplier Novus Biologicals Supplier No. NBP176646UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GRG (Groucho homolog) Polyclonal antibody specifically detects GRG (Groucho homolog) in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRG (Groucho homolog)
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias AES-1, AES-2, Amino enhancer of split, amino-terminal enhancer of split, ESP1, Gp130-associated protein GAM, GRG, GRG5, Protein ESP1, Protein GRG, TLE5
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the amino-terminus of human AES. The immunogen is located within the first 50 amino acids of AES. Amino Acid Squence: LQKRKLRLSKNHRPAR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 166
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.