missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35633-100ul
This item is not returnable.
View return policy
Description
GRF2 Polyclonal antibody specifically detects GRF2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| GRF2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 | |
| C3GGRF2guanine nucleotide-releasing factor 2 (specific for crk proto-oncogene), CRK SH3-binding GNRP, DKFZp781P1719, Guanine nucleotide-releasing factor 2, Protein C3G, Rap guanine nucleotide exchange factor (GEF) 1, rap guanine nucleotide exchange factor 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 844-1095 of human GRF2 (NP_941372.1).,, Sequence:, ARGVAARPGTLHDFHSHEIAEQLTLLDAELFYKIEIPEVLLWAKEQNEEKSPNLTQFTEHFNNMSYWVRSIIMLQEKAQDRERLLLKFIKIMKHLRKLNNFNSYLAILSALDSAPIRRLEWQKQTSEGLAEYCTLIDSSSSFRAYRAALSEVEPPCIPYLGLILQDLTFVHLGNPDYIDGKVNFSKRWQQFNILDSMRCFQQAHYDMRRNDDIINFFNDFSDHLAEEALWELSLKIKPRNITRRKTDREEKT | |
| 100 μL | |
| Signal Transduction | |
| 2889 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction