missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRF-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538253
This item is not returnable.
View return policy
Description
GRF-1 (glucocorticoid receptor DNA-binding factor 1), also known as p190RhoGAP or simply p190, is a transcriptional regulator which binds to the promoter region of the glucocorticoid receptor gene and represses its expression. By repressing GR expression, GRF-1 acts to down-regulate Rho signaling, thereby mediating both actin cytoskeletal rearrangements and cell cycle events. Through its GAP domain, GRF-1 is thought to affect cytokinesis by regulating Rho activity; a regulation that is controlled by the ubiquination of the GTP binding region and subsequent degradation of GRF-1. Additionally, GRF-1 plays an important role in oligodendrocyte differentiation, a process that is absent in maligt glioma tumors, implicating GRF-1 as a possible tumor suppressor. GRF-1 expression is regulated by glucocorticoids and the expressed protein exists as two isoforms produced by alternative splicing events.
Specifications
| GRF-1 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP35 | |
| Rho GTPase-activating protein 35, Glucocorticoid receptor DNA-binding factor 1, Glucocorticoid receptor repression factor 1, GRF-1, Rho GAP p190A, p190-A, ARHGAP35, GRF1, GRLF1, KIAA1722 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 232906, 2909, 306400 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NRY4, Q91YM2, P81128 | |
| ARHGAP35 | |
| A synthetic peptide derived from the internal region of human GRF-1 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction