Learn More
Manutan Grey medium duty steel shelving extension bay, 2000mm x 1000mm x 700mm
Product Code. 12835948
Encompass_Preferred
Product Code. 12835948 Supplier Manutan Supplier No. A360316
Only null left
Add to basket
Add to basket
Versatile add-on system that allows you to create a run of shelving as long as you require. All steel shelving for durability with easy boltless assembly. Shelves equipped with welded hooks and perforated every 62. 5mm for shelf adjustment. All components are powder coated for extra durability.
  • Shelf capacity UDL: 125kg
  • Dimensions (H x W x D): 2000mm x 1000mm x 700mm