missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Gremlin 1 Antibody (UCB patent anti-Gremlin-1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28165-1mg
This item is not returnable.
View return policy
Description
Gremlin 1 Monoclonal antibody specifically detects Gremlin 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Gremlin 1 | |
| Monoclonal | |
| Unconjugated | |
| O60565 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| UCB patent anti-Gremlin-1 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Cell proliferation-inducing gene 2 protein, CKTSF1B1gremlin 1, cysteine knot superfamily, homolog, Cysteine knot superfamily 1, BMP antagonist 1gremlin 1, cysteine knot superfamily, homolog (Xenopus laevis), DAN domain family member 2, DAND2GREMLIN, Down-regulated in Mos-transformed cells protein, DRMMGC126660, gremlin, gremlin 1, gremlin 1-like protein, gremlin-1, IHG-2, Increased in high glucose protein 2, increased in high glucose-2, proliferation-inducing gene 2 | |
| GREM1/ Gremlin | |
| 1 mg | |
| Neuroscience | |
| 26585 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction