missing translation for 'onlineSavingsMsg'
Learn More

Gremlin 1 Antibody (117), DyLight 550, Novus Biologicals™

Product Code. 18343650 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343650 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343650 Supplier Novus Biologicals Supplier No. NBP290353R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Gremlin 1 Monoclonal specifically detects Gremlin 1 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gremlin 1
Applications ELISA
Classification Monoclonal
Clone 117
Conjugate DyLight 550
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias Cell proliferation-inducing gene 2 protein, CKTSF1B1gremlin 1, cysteine knot superfamily, homolog, Cysteine knot superfamily 1, BMP antagonist 1gremlin 1, cysteine knot superfamily, homolog (Xenopus laevis), DAN domain family member 2, DAND2GREMLIN, Down-regulated in Mos-transformed cells protein, DRMMGC126660, gremlin, gremlin 1, gremlin 1-like protein, gremlin-1, IHG-2, Increased in high glucose protein 2, increased in high glucose-2, proliferation-inducing gene 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Gremlin 1 (O70326; Met1-Asp184).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 26585
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.