missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GREM2 Polyclonal Antibody

GREM2 Polyclonal Antibody
Brand: Invitrogen PA544813
This item is not returnable.
View return policy
Description
GREM2 Polyclonal Antibody for Western Blot
Specifications
| GREM2 | |
| Polyclonal | |
| Unconjugated | |
| Grem2 | |
| CKTSF1B2; CKTSF1B2FLJ21195; Cysteine knot superfamily 1, BMP antagonist 2; DAN domain family member 3; DAND3; DAND3Prdc; GREM2; gremlin 2; gremlin 2 homolog, cysteine knot superfamily; gremlin 2 homolog, cysteine knot superfamily (Xenopus laevis); gremlin 2, cysteine knot superfamily, homolog; gremlin 2, DAN family BMP antagonist; Gremlin1; Gremlin2; gremlin-2; Prdc; PRDCgremlin 2; Protein related to DAN and cerberus; RGD1560008 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64388 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H772 | |
| Grem2 | |
| Synthetic peptide directed towards the N-terminal of human GREM2 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction