missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GREM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119878
This item is not returnable.
View return policy
Description
Positive Control: CAL62 cell lysates, SH-SY5Y. Subcellular Location: Secreted.
GREM2 encodes a member of the BMP (bone morphogenic protein) antagonist family. Like BMPs, BMP antagonists contain cystine knots and typically form homo- and heterodimers. The CAN (cerberus and dan) subfamily of BMP antagonists, to which this gene belongs, is characterized by a C-terminal cystine knot with an eight-membered ring. The antagonistic effect of the secreted glycosylated protein encoded by this gene is likely due to its direct binding to BMP proteins. As an antagonist of BMP, this gene may play a role in regulating organogenesis, body patterning, and tissue differentiation.
Specifications
| GREM2 | |
| Polyclonal | |
| Unconjugated | |
| Grem2 | |
| CKTSF1B2; CKTSF1B2FLJ21195; Cysteine knot superfamily 1, BMP antagonist 2; DAN domain family member 3; DAND3; DAND3Prdc; GREM2; gremlin 2; gremlin 2 homolog, cysteine knot superfamily; gremlin 2 homolog, cysteine knot superfamily (Xenopus laevis); gremlin 2, cysteine knot superfamily, homolog; gremlin 2, DAN family BMP antagonist; Gremlin1; Gremlin2; gremlin-2; Prdc; PRDCgremlin 2; Protein related to DAN and cerberus; RGD1560008 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 64388 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9H772 | |
| Grem2 | |
| Synthetic peptide within human PRDC aa 50-100. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction