missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GREM1 Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543791
This item is not returnable.
View return policy
Description
In Xenopus, Gremlin belongs to a novel gene family that act as BMP antagonists in embryonic explants. The human homolog of Drm/Gremlin (CKTFS1B1) is localized on human chromosome 15q13--> q15. DRM-specific mRNA is expressed in different human tissues, including brain, ovary, intestine and colon. It has been suggested that down-regulation of DRM is associated with tumor progression, and support the hypothesis that human DRM may play an important role during both neuroembryological development and carcinogenesis.
Specifications
| GREM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O60565 | |
| Grem1 | |
| Lys26-Asp184 (Accession O60565), with N-terminal His Tag. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| C1 | |
| Unconjugated | |
| Grem1 | |
| C15DUPq; Cell proliferation-inducing gene 2 protein; CKTSF1B1; colorectal adenoma and carcinoma 1; CRAC1; CRCS4; Cysteine knot superfamily 1, BMP antagonist 1; DAN domain family member 2; DAND2; Down-regulated in Mos-transformed cells protein; down-regulated in v-mos-transformed cells; Drm; DUP15q; Grem; GREM1; gremlin; gremlin 1; gremlin 1 homolog, cysteine knot superfamily; gremlin 1, cysteine knot superfamily, homolog; gremlin 1, DAN family BMP antagonist; gremlin 1-like protein; gremlin-1; HMPS; HMPS1; IHG-2; Increased in high glucose protein 2; increased in high glucose-2; ld; MPSH; PIG2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 26585 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction