missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GREM1 Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

£397.00 - £1002.00

Specifications

Antigen GREM1 Biosimilar
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30284079
GREENER_CHOICE
View Documents
Invitrogen™
MA558609
100 μg
£397.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30284183
View Documents
Invitrogen™
MA558610
1 mg
£1002.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term UCB patent anti-Gremlin-1.

In Xenopus, Gremlin belongs to a novel gene family that act as BMP antagonists in embryonic explants. The human homolog of Drm/Gremlin (CKTFS1B1) is localized on human chromosome 15q13--> q15. DRM-specific mRNA is expressed in different human tissues, including brain, ovary, intestine and colon. It has been suggested that down-regulation of DRM is associated with tumor progression, and support the hypothesis that human DRM may play an important role during both neuroembryological development and carcinogenesis.
Specifications

Specifications

GREM1 Biosimilar
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.