Learn More
Greiner Bio-One Urine Collection Tubes
For collection and transport of urine specimens
Brand:  Greiner Bio-One 455003
Product Code. 15303701
  • £402.48 / Pack of 1200
Description
  • Sterile, leak-proof tubes made of clear, unbreakable disposable plastic
  • Pre-defined vacuum for exact draw volumes
  • Use tubes with round bottoms for chemical analyses; conical bottom tubes for microscopic examination of urine sediment
  • Use tubes with boric acid preservative for specimens tested for an otherwise unstable analyte or specimens for microbiological studies (stable up to 24 hours)
Tubes with preservatives should be inverted 6-8 times following collection in order to achieve homogenous mixing