missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Snap Caps
Click to view available options
Color:
Red
Unit Size:
Pack of 1000
Specifications
Specifications
| Product Type | Snap Cap |
| Material | PET |
| Color | Red |
| Size | Universal |
| For Use With (Equipment) | Recapping 13 mm Vacuette™ tubes to reduce the risk of biohazardous contamination |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction