missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One VACUETTE™ CAT Serum Clot Activator Blood Collection Tubes

Product Code. 12532936
Click to view available options
Capacity (Metric):
2 mL
4 mL
9 mL
Unit Size:
Pack of 1200
Pack of 50
This item is not returnable. View return policy

Product Code. 12532936

Brand: Greiner Bio-One 455092

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VACUETTE™ CAT Serum Clot Activator tubes, used for determinations in serum, are coated with micronized silica particles that activate clotting when the tubes are gently inverted.

  • VACUETTE™ Blood Collection Tubes are evacuated for easy, safe sample collection. The amount of vacuum in each tube corresponds to the tube’s blood volume specification and helps ensure the correct blood-to-additive ratio.
  • Injection molded from polyethylene terephthalate (PET) plastic, VACUETTE™ Blood Collection Tubes are highly transparent and virtually unbreakable.
  • VACUETTE™ Serum tubes include a clot-activating additive – micronized silica particles – that is spray-dried on the interior tube wall for even disbursement and optimal blood/additive mixing.
  • Optimal fill marks on every tube label indicate the proper draw volume for correct blood-to-additive ratios (within ±10% of the stated nominal volume in accordance with ISO and CLSI).
  • Featuring red VACUETTE™ SAFETY Caps, designed to minimize the spraying of aerosols during decapping and color-coded to aid first-glance tube recognition.
  • Sterile and latex-free.
TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Product Type Blood Collection Tube
Additive Clot Activator
Material PET
Sterility Sterile
Closure Type Non-ridged Pull Cap
Capacity (Metric) 9 mL
For Use With (Equipment) VACUETTE™ Blood Collection Needles
Diameter (Metric) 13 mm
Closure Color Red with Black Ring
Tube Size 16 x 100 mm
For Use With (Application) Blood Collection
Length (Metric) 100 mm
Diameter (Metric) Outer 16 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.